missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™
87.96 € - 147.27 €
Chemical Identifiers
CAS | 123-51-3 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
Synonym | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol |
PubChem CID | 31260 |
ChEBI | CHEBI:15837 |
IUPAC Name | 3-methylbutan-1-ol |
SMILES | CC(C)CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10205113
|
Fisher BioReagents
BP1754I-100 |
100 mL | Coated Amber Glass/Poison Pack | N/A | ||||||
10699543
|
Fisher BioReagents
BP1754I-400 |
400 mL | Coated Amber Glass/Poison Pack | N/A | ||||||
Description
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)
This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.Chemical Identifiers
123-51-3 | |
88.15 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CHEBI:15837 | |
CC(C)CCO |
C5H12O | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
Specifications
123-51-3 | |
15, 7354 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
≥99% | |
4.3 | |
Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture |
C5H12O | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CC(C)CCO | |
88.15 | |
CHEBI:15837 | |
Pass Test | |
Liquid |
Safety and Handling

missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : 4°C
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™