missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 6.7/8.0, Liq.), Fisher BioReagents™

Product Code. 10306413
Click to view available options
Quantity:
400 mL
Packaging:
Coated Amber Glass/Poison Pack

For extraction of proteins from crude nucleic acid preparations

This mixture is used for extracting protein from crude nucleic acid preparations. Solution comes with a bottle of tris that should be used to equilibrate the pH.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Color Clear
Assay Percent Range ≥99.0
Density 1.280g/cm³
Percent Purity ≥99.0%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
pH 6.8 to 8.2
Quantity 400 mL
Physical Form Liquid
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Chloroform, Phenol,Isoamyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'