Learn More
Phenol:Chloroform:Isoamyl Alcohol (25:24:1), Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Nucleic acid extraction and purification
81.50 € - 154.80 €
Specifications
Chemical Name or Material | Phenol:Chloroform:Isoamyl Alcohol (25:24:1) |
---|---|
UN Number | UN2922 |
Grade | Molecular Biology |
Packaging | Plastic bottle |
Physical Form | Liquid |
Description
Nucleic acid extraction and purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenol:Chloroform:Isoamyl Alcohol (25:24:1) | |
Molecular Biology | |
Liquid |
UN2922 | |
Plastic bottle |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335-H341-H350-H361d-H372
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.