missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized, saturated with 100 mM Tris-EDTA to pH 8.0, for molecular, Thermo Scientific Chemicals

Phenol-Chloroform-Isoamyl alcohol mixture in the ratio of 25:24:1 (v/v/v), CAS # 136112-00-0, 67-66-3, 123-51-3, is a solution used in the purification of nucleic acids.

92.66 € - 1002.99 €

Specifications

Chemical Name or Material Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized, saturated with 100 mM Tris-EDTA to pH 8.0
Color Yellow to Orange
Name Note DNase, RNase and Protease Free
CAS 136112-00-0, 67-66-3, 123-51-3
Molecular Formula C6H6O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10308293
View Documents
Thermo Scientific Acros
327111000
100 mL Glass bottle
92.66 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10535691
View Documents
Thermo Scientific Acros
327115000
500 mL Glass bottle
300.50 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10649273
View Documents
Thermo Scientific Acros
327110025
2.5 L Glass bottle
1002.99 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • This solution contains a mixture of pure chloroform, isoamyl alcohol and phenol saturated with Tris-EDTA, with a stabilized pH of 8.0

Applications

  • Phenol-Chloroform-Isoamyl alcohol mixture solution (25:24:1) is used in the extraction of nucleic acids, DNA, and RNA
  • When nucleic acids are extracted with the Phenol-Chloroform-Isoamyl alcohol mixture solution, a clear component separation is achieved. The proteins present in the sample are denatured and removed (collected in the organic phase or in the interphase), while nucleic acids remain in the aqueous phase. The chloroform denatures proteins and facilitates the separation of the aqueous and organic phases, and the isoamyl alcohol reduces foaming during extraction.
  • It is also used to generate DNA minisatellite fingerprints
Specifications

Specifications

Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized, saturated with 100 mM Tris-EDTA to pH 8.0
DNase, RNase and Protease Free
C6H6O
2927
14, 7241
Molecular Biology
1.28
7.8 to 8.2 (25°C)
Yellow to Orange
136112-00-0, 67-66-3, 123-51-3
MFCD00133763
1.2800g/mL
94.11
Glass bottle
Stabilized
Solution
Videos
Safety and Handling

Safety and Handling

Suspected of causing genetic defects, Suspected of damaging the unborn child, Causes damage to organs through prolonged or repeated exposure, Causes severe skin burns and eye damage, May cause drowsiness or dizziness, Suspected of causing cancer, Repeated exposure may cause skin dryness or cracking, Toxic if swallowed, in contact with skin or if inhaled

Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Specific target organ toxicity after repeated exposure (category 1), Skin corrosion/irritation (category 1), Specific target organ toxicity after single exposure (category 3), Carcinogenicity (category 2), Acute toxicity (category 3)

Product Identifier
  • Phenol/Chloroform/Isoamyl alcohol (25:24:1), stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Chloroform, Phenol,Isoamyl alcohol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only