Learn More
Phenol, detached crystals, 99+%, Thermo Scientific Chemicals
Description
Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A, which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers, which is used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colorings and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A, which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers, which is used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colorings and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.
Solubility
Soluble in water, alcohol, chloroform, ether, benzene, glycerol, acetone, carbon disulfide and aqueous alkali hydroxides.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Specifications
CAS | 108-95-2 |
Melting Point | 40°C to 43°C |
Density | 1.071 |
Boiling Point | 181°C |
Flash Point | 79°C |
Odor | Sweet |
Molecular Formula | C6H6O |
MDL Number | MFCD00002143 |
Quantity | 2500 g |
UN Number | UN1671 |
Show More |
Safety and Handling
- Phenol
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Phenol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.