missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Detached Crystals, SLR, Extra Pure, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Phenol Detached Crystals, SLR, Extra Pure
Specifications
Specifications
Melting Point | 39°C |
Color | Colorless to White |
pH | 6 |
Boiling Point | 182°C |
Packaging | Plastic powder jar |
Quantity | 500 g |
Viscosity | 3.437 mPaS (50°C) |
Formula Weight | 94.11g/mol |
Vapor Pressure | 0.4mbar at 20°C |
Physical Form | Crystalline Solid |
Safety and Handling
Product Identifier
- Phenol
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction