missing translation for 'onlineSavingsMsg'
Learn More

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Fisher Chemical™

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Fisher Chemical, Quantity: 5g, CAS: 620-45-1, Green, Molecular Formula: C12H6Cl2NNaO2, Molecular Weight: 290.075, Packaging: Glass Jar

30.80 € - 71.50 €

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10497450
View Documents
Fisher Chemical
P/2390/43
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10305860
View Documents
Fisher Chemical
P/2390/44
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Phenol-Indo-2,6-Dichlorophenol Sodium Salt, Pure, Fisher Chemical, Quantity: 5g, CAS: 620-45-1, Green, Molecular Formula: C12H6Cl2NNaO2, Molecular Weight: 290.075, Packaging: Glass Jar

Specifications

Chemical Identifiers

620-45-1
290.07
FHLDWQLHDYCXKI-UHFFFAOYSA-M
23697355
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
C12H6Cl2NNaO2
MFCD00150014
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
CHEBI:948

Specifications

620-45-1
Powder/Solid
MFCD00150014
FHLDWQLHDYCXKI-UHFFFAOYSA-M
sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate
23697355
325.9701g/mol
Green
C12H6Cl2NNaO2
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
290.07
CHEBI:948
Glass Jar
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.