missing translation for 'onlineSavingsMsg'
Learn More

Phenol Liquified 80% (w/w) In Water, Pure, Fisher Chemical

Product Code. 10090280
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle

Specifications

CAS 108-95-2
Melting Point 40.5°C
Color Red
Packaging Plastic Coated Glass bottle
Molecular Formula C6H6O
MDL Number MFCD00002143
Quantity 2.5 L
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 996
Vapor Density 3.2
ChEBI CHEBI:15882
Formula Weight 94.0414g/mol
Vapor Pressure 1mmHg 40
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol liquified
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.