missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, ACS Reagent, Honeywell Fluka™

Product Code. 15663830
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, ACS reagent

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

UN Number NONH for all modes of transport
Beilstein 326470
Formula Weight 354.38g/mol
Grade ACS Reagent
Packaging Glass Bottle
Visual Transition Interval 6.8 to 8.2 (yellow to red)
Color Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Phenol Red
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'