missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, Pure, Indicator Grade, Fisher Chemical™
Click to view available options
Quantity:
5 g
25 g
500 g
Specifications
Specifications
Melting Point | 300°C |
IUPAC Name | 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Formula Weight | 354.38g/mol |
Packaging | Glass Jar |
Color | Brown/Red |
Quantity | 25 g |
Physical Form | Powder |
Safety and Handling
Product Identifier
- Phenol Red
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction