missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Phenol Red, sodium salt Product Code.: 10418120

Thermo Scientific Chemicals Phenol Red, sodium salt

Product Code. 10418120
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10418120

Brand: Thermo Scientific Chemicals 151440250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285.0°C
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 376.35
Grade Pure
Packaging Glass bottle
Specific Extinction 1600 (558nm, in buffer) min.
Visual Transition Interval pH 6.8 (yellow) to pH 8.2 (red)
Color Brown to Red
Quantity 25 g
Physical Form Amorphous Powder
Chemical Name or Material Phenol Red, sodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0384115 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number151440050,151440250,151441000
Lot NumberA0446168 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number151440050,151440250,151441000
Lot NumberA0441768 Certificate TypeCertificate of Analysis Date03/05/2022 Catalog Number151440050,151440250,151441000
Lot NumberA0439023 Certificate TypeCertificate of Analysis Date24/02/2022 Catalog Number151440050,151440250,151441000
Lot NumberA0434092 Certificate TypeCertificate of Analysis Date22/09/2021 Catalog Number151440050,151440250,151441000
compliance-icons
Product Identifier
  • Phenol Red, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phenol Red, sodium salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.