missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol, Saturated (pH 4.3, Liq.), Fisher BioReagents
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Description
Phenol saturated with Tris buffer is used for purifying RNA.
Specifications
Specifications
Assay Percent Range | ≥99.0 |
Density | 1.05g/cm³ |
Percent Purity | ≥99.0% |
Identification | Pass Test |
Packaging | Coated Amber Glass/Poison Pack |
pH | 4.3 |
Quantity | 100 mL |
Physical Form | Liquid |
Chemical Name or Material | Phenol, Saturated |
Safety and Handling
Product Identifier
- Phenol, Saturated (pH 4.3)
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- MIXTURE LIST-Contains : Phenol, 1,2-Benzenedicarboxylic acid, monopotassium salt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction