missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Saturated (pH 6.6/7.9, Liq.), Fisher BioReagents™

Product Code. 10264673
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack

Phenol, Saturated (pH 6.6/7.9, Liq.)

Phenol saturated with Tris buffer is used in DNA extraction procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2, 6381-92-6, 77-86-1
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Assay Percent Range ≥99.0
UN Number 2821
Percent Purity ≥99.0%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenol, Saturated
Grade DNase- and RNase-Free
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol saturated pH 6.6
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Phenol, Tris (hydroxymethyl) aminomethane, Ethylenediaminetetraacetic acid, disodium salt dihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'