missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol, ultrapure, 99%, unstab.

Product Code. 15466149
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.

Solubility
Soluble in water (82 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Color White
Density 1.071
Boiling Point 181°C
Flash Point 79°C (175°F)
Odor Sweet
Quantity 500 g
UN Number UN1671
Beilstein 969616
Sensitivity Air Sensitive
Merck Index 14,7241
Solubility Information Soluble in water (82g/L at 20°C).
Formula Weight 94.11
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Phenol, unstabilized
Grade Ultrapure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.