missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol USP Crystal, Macron Fine Chemicals™
Click to view available options
Quantity:
2.5 kg
12 x 500 g
Packaging:
Glass
Poly Coated
missing translation for 'unitSize'
2.5kg
500g
Especificaciones
Especificaciones
| Packaging | Poly Coated |
| Quantity | 2.5 kg |
| Formula Weight | 94.11 |
| Grade | USP |
| Physical Form | Crystalline |
| Chemical Name or Material | Phenol |
Seguridad y manipulación
Palabra de advertencia
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido