missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Solution 1% In Ethanol, Pure, Indicator Grade, Fisher Chemical™

C20H14O4, CAS Number-77-09-8, 500mL, CHEBI:34914, Yellow, KJFMBFZCATUALV-UHFFFAOYSA-N, 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one, 5913, 318.328, Amber glass bottle, Liquid, 4764, C1-CC-C2C(-C1)C(-O)OC2(C3-CC-C(C-C3)O)C4-CC-C(C-C4)O

Brand:  Fisher Chemical P/2401L/08

47.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10274200

  • 66.10 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

C20H14O4, CAS Number-77-09-8, 500mL, CHEBI:34914, Yellow, KJFMBFZCATUALV-UHFFFAOYSA-N, 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one, 5913, 318.328, Amber glass bottle, Liquid, 4764, C1-CC-C2C(-C1)C(-O)OC2(C3-CC-C(C-C3)O)C4-CC-C(C-C4)O

TRUSTED_SUSTAINABILITY
Specifications

Specifications

77-09-8
MFCD00005913
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
Amber glass bottle
500 mL
C20H14O4
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
Pure
Yellow
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phenolphthalein, 1% solution

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Specific target organ toxicity Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H371-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/A§/if you feel unwell.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.