missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol, Honeywell Fluka™
Click to view available options
Quantity:
30 mL
500 mL
1 L
50 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
UN Number | UN1170 |
Beilstein | 284423 |
IUPAC Name | 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one |
Formula Weight | 318.32g/mol |
Quantity | 6 x 1 L |
Flash Point | 14°C (57.2°F) |
Chemical Name or Material | Phenolphthalein solution |
Safety and Handling
Product Identifier
- Phenolphthalein solution
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : phenolphthalein, ethanol; ethyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction