missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol, Honeywell Fluka™

Product Code. 15698370
Click to view available options
Quantity:
30 mL
500 mL
1 L
50 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Indicator, Reag. Ph. Eur., 1% in ethanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

UN Number UN1170
Beilstein 284423
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Formula Weight 318.32g/mol
Quantity 30 mL
Flash Point 14°C (57.2°F)
Chemical Name or Material Phenolphthalein solution
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenolphthalein solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : phenolphthalein, ethanol; ethyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.