missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein TS/RS Solution According to United States Pharmacopoeia (USP), Reagecon
BIOPHARMA

Specified for use in individual United States Pharmacopoeia (USP) Monographs and tested and certified by an ISO17025 Accredited Test Method (INAB Ref. 264T).

Brand:  Reagecon USP4201


Product Code. 13499038

  • 185.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • United States Pharmacopoeia (USP) Reference Standard, United States Pharmacopoeia (USP) Reference Material
  • Specified for use in individual United States Pharmacopoeia (USP) Monographs.
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref. 264T).
  • NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Amber Glass Winchester Bottle
100 mL
Titration and USP Monographs
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • phenolphthalein TS

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol, phenolphthalein

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.