missing translation for 'onlineSavingsMsg'
Learn More

Phenosafranin, Thermo Scientific Chemicals

Product Code. 10496373
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 81-93-6
Melting Point >300.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C18H15ClN4
MDL Number MFCD00036335
Quantity 1 g
Merck Index 15,7363
Synonym phenosafranin, phenosafranine, phenosafranine, chloride, phenosafrinine, phenosofranine, phenosofrinine, safranin b extra, safrinin b extra, 3,7-diamino-5-phenylphenazinium chloride, schultz no. 958
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
InChI Key SOUHUMACVWVDME-UHFFFAOYSA-N
SMILES [Cl-].NC1=CC2=[N+](C3=CC=CC=C3)C3=CC(N)=CC=C3N=C2C=C1
IUPAC Name 10-phenylphenazin-10-ium-2,8-diamine;chloride
Molecular Weight (g/mol) 322.80
PubChem CID 65732
ChEBI CHEBI:33601
Formula Weight 322.8
Chemical Name or Material Phenosafranin
Show More Show Less
compliance-icons
Product Identifier
  • Phenosafranin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.