missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenothiazine, 98+%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
500 g
5000 g
Description
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.
Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.
Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
Specifications
Specifications
Melting Point | 182°C to 186°C |
Boiling Point | 235°C (21 mmHg) |
Quantity | 500 g |
Beilstein | 143237 |
Sensitivity | Light sensitive |
Merck Index | 14,7252 |
Solubility Information | Soluble in water (0.13mg/ml at 23°C),ethanol,acetone,and ether. Insoluble in chloroform. |
Formula Weight | 199.28 |
Percent Purity | ≥98% |
Chemical Name or Material | Phenothiazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'