missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10572381
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-99-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000726
InChI Key PKUPAJQAJXVUEK-UHFFFAOYSA-N
Synonym phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride
PubChem CID 69703
IUPAC Name 2-phenoxyacetyl chloride
SMILES ClC(=O)COC1=CC=CC=C1

Specifications

Color Yellow to Brown
Density 1.2300g/mL
Boiling Point 225°C to 226°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5OCH2COCl
Refractive Index 1.5324 to 1.5344
Quantity 50 g
Beilstein 06, 162
Specific Gravity 1.23
Solubility Information Solubility in water: reacts
Formula Weight 170.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Phenoxyacetyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenoxyacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'