missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl acrylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Density | 1.076 |
Boiling Point | 89°C to 90°C (12 mmHg) |
Refractive Index | 1.521 |
Quantity | 25 g |
UN Number | UN3082 |
Beilstein | 2041124 |
Solubility Information | Insoluble in water. |
Formula Weight | 148.16 |
Percent Purity | 97% |
Chemical Name or Material | Phenyl acrylate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phenyl acrylate, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction