missing translation for 'onlineSavingsMsg'
Learn More

Phenyl benzoate, 99%, Thermo Scientific Chemicals

Product Code. 10695212
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-99-2
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00003072
InChI Key FCJSHPDYVMKCHI-UHFFFAOYSA-N
Synonym benzoic acid phenyl ester, diphenylcarboxylate, benzoic acid, phenyl ester, benzoic acid phenyl, phenylbenzoate, unii-b8a3wvz590, phenol, benzoate, dsstox_cid_28185, dsstox_rid_82744, dsstox_gsid_48210
PubChem CID 7169
ChEBI CHEBI:86919
IUPAC Name phenyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2

Specifications

Acidity 1mg KOH/g max.
Melting Point 68.0°C to 70.0°C
Color White
Boiling Point 298.0°C to 299.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5CO2C6H5
Quantity 500 g
Beilstein 09, 116
Merck Index 15, 7388
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and chloroform
Formula Weight 198.22
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenyl benzoate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'