missing translation for 'onlineSavingsMsg'
Learn More
Phenyl disulfide, 99%, Thermo Scientific Chemicals Product Code.: 10052050

Phenyl disulfide, 99%, Thermo Scientific Chemicals

Product Code. 10052050
50 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10052050

Brand: Thermo Scientific Acros 173710500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.33
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2

Specifications

Melting Point 58°C to 61°C
Color White
Boiling Point 191°C to 192°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5S-)2
Quantity 50 g
Beilstein 06, 323
Fieser 05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156
Merck Index 15, 3355
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 218.33
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Phenyl disulfide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenyl disulfide
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenyl disulfide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.