missing translation for 'onlineSavingsMsg'
Learn More

Phenyl disulfide, 99%, Thermo Scientific Chemicals

Product Code. 15790819
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.33
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2

Specifications

Melting Point 58°C to 61°C
Color White
Boiling Point 191°C to 192°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5S-)2
Quantity 250 g
Beilstein 06, 323
Fieser 05,276; 06,235; 08,210; 10,171; 11,220; 12,514; 13,156
Merck Index 15, 3355
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
Formula Weight 218.33
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Phenyl disulfide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'