missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl ether, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 26.0°C to 30.0°C |
Density | 1.0730g/mL |
Boiling Point | 259.0°C |
Flash Point | 115°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (C6H5)2O |
Refractive Index | 1.5795 to 1.5815 |
Quantity | 500 g |
Beilstein | 06,146 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0448250 | Certificate of Analysis | 31/01/2025 | 130605000,130601000,130600010,130600100,130600025 |
A0467070 | Certificate of Analysis | 31/01/2025 | 130605000,130601000,130600010,130600100,130600025 |
A0448250 | Certificate of Analysis | 25/12/2023 | 130605000,130601000,130600010,130600100,130600025 |
A0442541 | Certificate of Analysis | 25/08/2023 | 130605000,130601000,130600010,130600100,130600025 |
A0397527 | Certificate of Analysis | 15/12/2022 | 130605000,130601000,130600010,130600100,130600025 |
Safety and Handling


Product Identifier
- Phenyl ether
- Warning
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phenyl ether, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction