missing translation for 'onlineSavingsMsg'
Learn More

Phenyl-p-benzoquinone, 95%, Thermo Scientific Chemicals

123.00 € - 312.00 €

Chemical Identifiers

CAS 363-03-1
Molecular Formula C12H8O2
Molecular Weight (g/mol) 184.19
MDL Number MFCD00001599
InChI Key RLQZIECDMISZHS-UHFFFAOYSA-N
Synonym phenyl-p-benzoquinone, phenylquinone, phenylbenzoquinone, 2-phenyl-1,4-benzoquinone, 2-phenylbenzoquinone, p-benzoquinone, 2-phenyl, 2,5-cyclohexadiene-1,4-dione, 2-phenyl, 2-phenyl-p-benzoquinone, phenyl-1,4-benzoquinone, p-benzoquinone, phenyl
PubChem CID 9688
IUPAC Name 2-phenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=CC(=O)C(=C1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10445471
View Documents
Thermo Scientific Acros
25 g Glass bottle
123.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
10331261
View Documents
Thermo Scientific Acros
417381000
100 g Glass bottle
312.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

363-03-1
184.19
RLQZIECDMISZHS-UHFFFAOYSA-N
9688
O=C1C=CC(=O)C(=C1)C1=CC=CC=C1
C12H8O2
MFCD00001599
phenyl-p-benzoquinone, phenylquinone, phenylbenzoquinone, 2-phenyl-1,4-benzoquinone, 2-phenylbenzoquinone, p-benzoquinone, 2-phenyl, 2,5-cyclohexadiene-1,4-dione, 2-phenyl, 2-phenyl-p-benzoquinone, phenyl-1,4-benzoquinone, p-benzoquinone, phenyl
2-phenylcyclohexa-2,5-diene-1,4-dione

Specifications

363-03-1
Brown-Yellow
94% min. (HPLC)
MFCD00001599
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,benzene,petroleum ether;,very soluble in chloroform
O=C1C=CC(=O)C(=C1)C1=CC=CC=C1
184.19
184.19
Chunks and/or Powder
111°C to 115°C
Authentic
C12H8O2
phenyl-p-benzoquinone, phenylquinone, phenylbenzoquinone, 2-phenyl-1,4-benzoquinone, 2-phenylbenzoquinone, p-benzoquinone, 2-phenyl, 2,5-cyclohexadiene-1,4-dione, 2-phenyl, 2-phenyl-p-benzoquinone, phenyl-1,4-benzoquinone, p-benzoquinone, phenyl
RLQZIECDMISZHS-UHFFFAOYSA-N
2-phenylcyclohexa-2,5-diene-1,4-dione
9688
95%
Phenyl-p-benzoquinone, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 206-654-5

missing translation for 'rtecsNumber' : DK6650000

missing translation for 'tsca' : TSCA

Product Identifier
  • Phenyl-p-benzoquinone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only