missing translation for 'onlineSavingsMsg'
Learn More

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 10184690
10 g, Glass bottle
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66778-08-3
Molecular Formula C6H5Na2O42H2O
Molecular Weight (g/mol) 254.09
MDL Number MFCD00149080
InChI Key UMRLIPYPVZYYFT-UHFFFAOYSA-N
Synonym sodiumphenylphosphatedibasicdihydrate
PubChem CID 131883070
IUPAC Name phenyl dihydrogen phosphate;sodium;dihydrate
SMILES C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5OP(O)(ONa)2·2H2O
Quantity 10 g
Merck Index 14, 3357
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether
Formula Weight 254.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Phenyl phosphate, disodium salt dihydrate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.