missing translation for 'onlineSavingsMsg'
Learn More

Phenyl propargyl ether, 97%, Thermo Scientific Chemicals

36.38 € - 561.00 €

Chemical Identifiers

CAS 13610-02-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00048107
InChI Key AIQRJSXKXVZCJO-UHFFFAOYSA-N
Synonym phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
PubChem CID 83613
IUPAC Name prop-2-ynoxybenzene
SMILES C#CCOC1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043923.03
1 g
36.38 €
1g
Please to purchase this item. Need a web account? Register with us today!
11304978
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043923.06
5 g
57.18 €
5g
Please to purchase this item. Need a web account? Register with us today!
11394968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043923.14
25 g
164.82 €
25g
Please to purchase this item. Need a web account? Register with us today!
11314978
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043923.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Phenyl propargyl ether is used as building block for synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13610-02-1
132.16
AIQRJSXKXVZCJO-UHFFFAOYSA-N
83613
C#CCOC1=CC=CC=C1
C9H8O
MFCD00048107
phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
prop-2-ynoxybenzene

Specifications

13610-02-1
1.030 g/mL
Pungent
C6H5OCH2C≡CH
MFCD00048107
Soluble in chloroform.
C#CCOC1=CC=CC=C1
132.16
132.16
Liquid
Colorless to Yellow
89°C to 90°C (14 mmHg)
C9H8O
1.533
phenyl propargyl ether, prop-2-ynyloxy benzene, prop-2-yn-1-yloxy benzene, phenyl 2-propynyl ether, benzene, 2-propynyloxy, 3-phenoxy-1-propyne, 2-propynyloxy benzene, phenylpropargyl ether, prop-2-ynyloxybenzene, ether, phenyl 2-propynyl
AIQRJSXKXVZCJO-UHFFFAOYSA-N
prop-2-ynoxybenzene
83613
97%
Phenyl propargyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H318-H335

missing translation for 'einecsNumber' : 237-095-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only