Learn More
Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media
Used for capture and intermediate purification requiring low to medium hydrophobicity
Brand:  Cytiva 17-0965-10
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10231400
  • 192.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Base matrix derivatized via uncharged, chemically-stable ether linkages
  • Two levels of substitution help find optimal selectivity and binding capacity for given application
  • Hydrophilic nature of the base matrix ensures low levels of non-specific binding
  • Hydrophobic interaction chromatography (HIC) media composed of cross-linked 6% agarose beads modified with standard aromatic phenyl groups via uncharged, chemically-stable ether linkages
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Degree of substitution for low sub HIC media is ∽25μmol phenyl/mL gel
  • Degree of substitution for high sub HIC media is ∽40μmol phenyl/mL gel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
For Capture and Intermediate PurifiCatIon Requiring Low to Medium Hydrophobicity
Phenyl Sepharose 6 Fast Flow Media (low sub)
45 to 165 μm
3 to 13
Phenyl
Stable in commonly used aqueous Buffers: 1 M NaOH, 3 M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6 M Guanidine-Hydrochloride, 8 M Urea
4°C to 30°C
250 to 400 cm/Hoμr
6% Cross-Linked Agarose
20% Ethanol
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Phenyl Sepharose 6 Fast Flow (low sub)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol