missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Phenyl Sepharose™ CL-4B Gel
For immunoglobulin purification and fractionation as well as immunoprecipitation procedures
Brand: Cytiva 17-0810-01
This item is not returnable.
View return policy
Description
- Strong binding to Fc region of IgG
- Excellent for polyclonal antibody purification
- High capacity: 20mg human IgG/mL
Specifications
Phenyl Sepharose CL-4 B | |
For Hydrophobic Interaction Chromato Graphy | |
Phenyl | |
3 to 12 | |
4°C to 30°C, 20% Ethanol or 0.1 M NaOH |
<150 cm/hr | |
200 mL | |
45 μm-165 μm | |
4% Cross-linked Agarose |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Phenyl Sepharose CL-4B, 200 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol