missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Specifications
Specifications
Acidity | 1mg KOH/g max. |
Color | Colorless to Yellow |
Density | 1.004g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C10H14O2 |
Linear Formula | C6H5CH2CH(OCH3)2 |
Refractive Index | 1.4930 to 1.4950 (20°C, 589nm) |
Quantity | 500 g |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Safety and Handling
Product Identifier
- Phenylacetaldehyde dimethyl acetal
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Phenylacetaldehyde dimethyl acetal, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction