missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic acid hydrazide, 98%, Thermo Scientific Chemicals

Product Code. 10264750
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-39-3
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00007612
InChI Key FPTCVTJCJMVIDV-UHFFFAOYSA-N
Synonym phenylacetic acid hydrazide, benzeneacetic acid, hydrazide, phenylacetic hydrazide, phenylacetylhydrazine, phenylacetyl hydrazide, phenacetic acid hydrazide, 2-phenylacetyl hydrazine, phenylacetohydrazide, phenylacethydrazide, phenyl-acetic acid hydrazide
PubChem CID 70301
IUPAC Name 2-phenylacetohydrazide
SMILES C1=CC=C(C=C1)CC(=O)NN

Specifications

Melting Point 115.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CONHNH2
Quantity 25 g
Beilstein 09, 446
Formula Weight 150.18
Percent Purity 98%
Physical Form Needles
Chemical Name or Material Phenylacetic acid hydrazide
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetic acid hydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.