missing translation for 'onlineSavingsMsg'
Learn More

Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals

54.87 € - 513.56 €

Chemical Identifiers

CAS 98-80-6
Molecular Formula C6H7BO2
Molecular Weight (g/mol) 121.93
MDL Number MFCD00002103
InChI Key HXITXNWTGFUOAU-UHFFFAOYSA-N
Synonym benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
PubChem CID 66827
ChEBI CHEBI:44923
IUPAC Name phenylboronic acid
SMILES OB(O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10041420
View Documents Promotion Details
Thermo Scientific Acros
130360100
10 g Glass bottle
54.87 €
10g
Please to purchase this item. Need a web account? Register with us today!
10667672
View Documents Promotion Details
Thermo Scientific Acros
130360500
50 g Glass bottle
173.87 €
50g
Please to purchase this item. Need a web account? Register with us today!
10440194
View Documents Promotion Details
Thermo Scientific Acros
130362500
250 g Glass bottle
513.56 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-80-6
121.93
HXITXNWTGFUOAU-UHFFFAOYSA-N
66827
phenylboronic acid
C6H7BO2
MFCD00002103
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
CHEBI:44923
OB(O)C1=CC=CC=C1

Specifications

98-80-6
100.0
White
98% min. (HPLC)
C6H5B(OH)2
16, 920
15, 1070
Solubility in water: 10g/L (20°C). Other solubilities: solubility in benzene : 1.75%, xylene : 1.2%, methanol : 178%, ether : 30.2%
OB(O)C1=CC=CC=C1
121.93
CHEBI:44923
98+%
Phenylboronic acid
96.5
217°C to 222°C
Authentic
C6H7BO2
MFCD00002103
01,833; 02,317; 03,221; 05,513; 07,284; 09,23; 11,408; 15,254
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2
HXITXNWTGFUOAU-UHFFFAOYSA-N
phenylboronic acid
66827
121.93
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 202-701-9

Product Identifier
  • Phenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Boronic acid, phenyl-

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only