missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 24388-23-6 |
Melting Point | 27°C to 31°C |
Boiling Point | 110°C (12.0 Torr) |
Flash Point | 107°C |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C12H17BO2 |
MDL Number | MFCD00966985 MFCD31699955 |
Quantity | 25 g |
Synonym | phenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, phenylboronic acid, pinacol ester, 2-phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzeneboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenyl, acmc-209gbq, phenylboronic pinacol ester, phenylboronicacidpinacolester, pinacolboryl benzene |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'