missing translation for 'onlineSavingsMsg'
Learn More

Phenylhydrazine, 97%, Thermo Scientific Chemicals

Product Code. 11915661
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-63-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007573
InChI Key HKOOXMFOFWEVGF-UHFFFAOYSA-N
Synonym hydrazinobenzene, hydrazine, phenyl, phenylhydrazin, monophenylhydrazine, phenyl hydrazine, 1-phenylhydrazine, hydrazine-benzene, fenilidrazina, fenylhydrazine, fenylhydrazine dutch
PubChem CID 7516
ChEBI CHEBI:27924
IUPAC Name phenylhydrazine
SMILES C1=CC=C(C=C1)NN

Specifications

Melting Point 19.0°C
Color Beige or Blue to White
Density 1.0900g/mL
Boiling Point 238.0°C to 241.0°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5NHNH2
Refractive Index 1.6040 to 1.6090
Quantity 2.5 kg
Beilstein 15,II,44
Fieser 01,838; 02,322; 06,457; 07,284
Merck Index 15,7404
Specific Gravity 1.09
Solubility Information Solubility in water: 145g/L (20°C). Other solubilities: soluble in diluted acids,miscible with alcohol,ether,chloroform and,benzene,sparingly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Phenylhydrazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'