missing translation for 'onlineSavingsMsg'
Learn More

Phenylmagnesium bromide, 3M in 2-MeTHF, Thermo Scientific Chemicals

Product Code. 15410938
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Phenyl magnesium bromide is the suitable reagent used for the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s. It may be used for synthesis of  1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol, (3-(2-Dithiobenzoatepropionyl)propyl)dimethylmethoxysilane, reversible addition-fragmentation chain transfer polymerization (RAFT)-silane agent, series of o-substituted benzophenones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl magnesium bromide is the suitable reagent used for the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s. It may be used for synthesis of  1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, 6-benzyl-1,
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-58-3
Molecular Formula C6H5BrMg
Linear Formula C6H5MgBr
MDL Number MFCD00000038
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym phenylmagnesium bromide, bromophenylmagnesium, magnesium, bromophenyl, phmgbr, phenylmagnesiumbromide, bromo phenyl magnesium, phenyl magnesium bromide, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, grignard reagent
Solubility Information Reacts with water.
InChI Key ANRQGKOBLBYXFM-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=CC=C1
IUPAC Name magnesium;benzene;bromide
Molecular Weight (g/mol) 181.32
PubChem CID 66852
Formula Weight 181.31
Concentration or Composition (by Analyte or Components) 3M in 2-MeTHF
Chemical Name or Material Phenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.