missing translation for 'onlineSavingsMsg'
Learn More
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals Product Code.: 10502151

Phenylphosphonic acid, 98%, Thermo Scientific Chemicals

Product Code. 10502151
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10502151

Brand: Thermo Scientific Acros 130791000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-33-1
Molecular Formula C6H7O3P
Molecular Weight (g/mol) 158.09
MDL Number MFCD00002136
InChI Key QLZHNIAADXEJJP-UHFFFAOYSA-N
Synonym benzenephosphonic acid, phosphonic acid, phenyl, phenyl-phosphonic acid, phosphonic acid, p-phenyl, phenyl phosphonic acid, unii-byd76t2868, sv7, pubchem21303, phosphonic acid,phenyl, wln: qpqo&rShow MoreShow Less
PubChem CID 15295
IUPAC Name phenylphosphonic acid
SMILES OP(O)(=O)C1=CC=CC=C1

Specifications

Melting Point 158.0°C to 165.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5P(O)(OH)2
Quantity 100 g
Beilstein 16,803
Solubility Information Solubility in water: 40.4g/100mL water (25°C). Other solubilities: soluble in alcohol and ccl4
Formula Weight 158.09
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Phenylphosphonic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458439 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number130790025,130791000,130795000
Lot NumberA0400227 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number
Lot NumberA0454778 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number130790025,130791000,130795000
Lot NumberA0393326 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number
Lot NumberA0394855 Certificate TypeCertificate of Analysis Date10/09/2020 Catalog Number
compliance-icons compliance-icons
Product Identifier
  • Phenylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.