missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11499006
Click to view available options
Quantity:
5 g
25 g
100 g

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases), and most common organic solvents.

Notes
Moisture Sensitive. Store away from water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-53-1
Molecular Formula C6 H8 Si
Molecular Weight (g/mol) 108.22
MDL Number MFCD00013478
InChI Key XJWOWXZSFTXJEX-UHFFFAOYSA-N
Synonym phenylsilane, benzene, silyl, silylbenzene, silane, phenyl, fenylsilan, fenylsilan czech, phenylsilane, silyl, silane, phenyl-,
PubChem CID 6327628
IUPAC Name phenylsilicon
SMILES C1=CC=C(C=C1)[Si]

Specifications

Boiling Point 120°C
Quantity 5 g
UN Number UN3399
Flash Point 7°C (45°F)
Solubility Information Contact with water releases a flammable alcohol. Soluble in water (Contact with water releases flammable gases),and most common organic solvents.
Formula Weight 108.22
Percent Purity 97%
Refractive Index 1.5105
Sensitivity Moisture sensitive
Density 0.88
Chemical Name or Material Phenylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phenylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Phenylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.