missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals

Product Code. 11443994
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-51-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004256
InChI Key LVFFZQQWIZURIO-UHFFFAOYSA-N
Synonym phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
PubChem CID 95459
IUPAC Name 2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O

Specifications

Melting Point 167°C to 169°C
Quantity 25 g
Beilstein 1875051
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material (±)-Phenylsuccinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'