missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals

Product Code. 11463994
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-51-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004256
InChI Key LVFFZQQWIZURIO-UHFFFAOYSA-N
Synonym phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
PubChem CID 95459
IUPAC Name 2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O

Specifications

Melting Point 167°C to 169°C
Quantity 500 g
Beilstein 1875051
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material (±)-Phenylsuccinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.