Learn More
Phenyltrimethoxysilane, 97%, Thermo Scientific Chemicals
27.30 € - 185.00 €
Chemical Identifiers
CAS | 2996-92-1 |
---|---|
Molecular Formula | C9H14O3Si |
Molecular Weight (g/mol) | 198.29 |
MDL Number | MFCD00025689 |
InChI Key | ZNOCGWVLWPVKAO-UHFFFAOYSA-N |
Synonym | phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane |
PubChem CID | 18137 |
IUPAC Name | trimethoxy(phenyl)silane |
SMILES | CO[Si](OC)(OC)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494914
|
Thermo Scientific Alfa Aesar
A13827.14 |
25 g |
27.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404924
|
Thermo Scientific Alfa Aesar
A13827.22 |
100 g |
50.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414924
|
Thermo Scientific Alfa Aesar
A13827.36 |
500 g |
185.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
Chemical Identifiers
2996-92-1 | |
198.29 | |
ZNOCGWVLWPVKAO-UHFFFAOYSA-N | |
18137 | |
CO[Si](OC)(OC)C1=CC=CC=C1 |
C9H14O3Si | |
MFCD00025689 | |
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane | |
trimethoxy(phenyl)silane |
Specifications
210°C to 212°C | |
2996-92-1 | |
MFCD00025689 | |
86°C (186°F) | |
Reacts with water. | |
CO[Si](OC)(OC)C1=CC=CC=C1 | |
198.29 | |
198.3 | |
1.4735 | |
1.07 |
-25°C | |
C9H14O3Si | |
2937896 | |
phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane | |
ZNOCGWVLWPVKAO-UHFFFAOYSA-N | |
trimethoxy(phenyl)silane | |
18137 | |
97% | |
Moisture sensitive | |
Phenyltrimethoxysilane |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P314-P330-P370+P378q-P501c
H226-H302-H373
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 221-066-9
missing translation for 'rtecsNumber' : VV5252000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only