Learn More
Phenyltrimethylsilane, 98%, Thermo Scientific Chemicals
30.35 € - 82.10 €
Chemical Identifiers
CAS | 768-32-1 |
---|---|
Molecular Formula | C9H14Si |
Molecular Weight (g/mol) | 150.30 |
MDL Number | MFCD00008271 |
InChI Key | KXFSUVJPEQYUGN-UHFFFAOYSA-N |
Synonym | phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane |
PubChem CID | 69849 |
IUPAC Name | trimethyl(phenyl)silane |
SMILES | C[Si](C)(C)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397875
|
Thermo Scientific Alfa Aesar
L02876.06 |
5 g |
30.35 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307885
|
Thermo Scientific Alfa Aesar
L02876.14 |
25 g |
82.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as intermediate for pharmaceutical and organic synthesis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
768-32-1 | |
150.30 | |
KXFSUVJPEQYUGN-UHFFFAOYSA-N | |
69849 | |
C[Si](C)(C)C1=CC=CC=C1 |
C9H14Si | |
MFCD00008271 | |
phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane | |
trimethyl(phenyl)silane |
Specifications
169°C to 170°C | |
C9H14Si | |
UN1993 | |
40°C (104°F) | |
Not miscible or difficult to mix with water. | |
C[Si](C)(C)C1=CC=CC=C1 | |
150.30 | |
150.3 | |
1.49 | |
Phenyltrimethylsilane |
768-32-1 | |
MFCD00008271 | |
1855012 | |
phenyltrimethylsilane, trimethyl phenyl silane, silane, trimethylphenyl, trimethylsilyl benzene, unii-px1sg1q5yp, benzene, trimethylsilyl, silane, trimethyl phenyl, px1sg1q5yp, phenyl trimethylsilane, trimethyl-phenylsilane | |
KXFSUVJPEQYUGN-UHFFFAOYSA-N | |
trimethyl(phenyl)silane | |
69849 | |
98% | |
0.871 |
Safety and Handling
GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 212-192-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only