missing translation for 'onlineSavingsMsg'
Learn More

Phenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15415355
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as primary and secondary intermediate.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids a
TRUSTED_SUSTAINABILITY

Specifications

CAS 23665-09-0
Density 0.966
Molecular Formula C6H5IZn
MDL Number MFCD00671995
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym phenylzinc iodide, zinc, iodophenyl, benzene,iodozinc 1+, iodo phenyl zinc, phenylzinc iodide, 0.50 m in thf, phenylzinc iodide 0.5 m in tetrahydrofuran, phenylzinc iodide solution, 0.5 m in thf, phenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key QXXJGZQLPDQOPF-UHFFFAOYSA-M
SMILES C1=CC=[C-]C=C1.[Zn+]I
IUPAC Name benzene;iodozinc(1+)
Molecular Weight (g/mol) 269.39
PubChem CID 4117658
Formula Weight 269.39
Chemical Name or Material Phenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.