Learn More
uvex Pheos B-S-WR Safety Helmet
Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.
454.00 €
Specifications
Certifications/Compliance EN 397
For Use With (Equipment) Hard Hats
Material HDPE
Model 9772
Product Type Industrial Safety Helmet
View More Specs
Products 1
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17514116
View Documents
uvex
9772031
White
454.00 €
Pack of 25
Estimated Shipment: 06-01-2025
to see stock.
Add to basket
Description
Description
  • Lightweight helmet in a sporty design
  • Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • Shorter brim for a wider field of upward vision
  • 6-point textile suspension harness with rotating size adjusters for individual width adjustment
Specifications
Specifications
EN 397
HDPE
Industrial Safety Helmet
52 to 61 cm
Ventilated
Hard Hats
9772
52 to 61 cm
Short Brim
Front Brim