missing translation for 'onlineSavingsMsg'
Learn More

uvex Pheos E Safety Helmet

Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.

294.00 €

Specifications

Certifications/Compliance EN 397, EN 50365
For Use With (Equipment) Hard Hats
Material HDPE
Model 9770
Product Type Industrial Safety Helmet
View More Specs

Products 1
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17554106
View Documents
uvex
9770020
White
294.00 €
Pack of 25
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

  • Lightweight helmet in a sporty design
  • Helmet shell fully closed for use by electricians
  • Tested and certified in accordance with EN 397 and EN 50365 (1000 VAC)
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • Conventional 6-point textile suspension harness for a comfortable fit
Specifications

Specifications

EN 397, EN 50365
HDPE
Industrial Safety Helmet
51 to 61 cm
Not Ventilated
Hard Hats
9770
52 to 61 cm
Long Brim
Front Brim
Videos
SDS
Documents

Documents

Product Certifications