missing translation for 'onlineSavingsMsg'
Learn More

uvex Pheos E-WR Safety Helmet

Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.

420.00 €

Specifications

Certifications/Compliance EN 397, EN 50365
For Use With (Equipment) Hard Hats
Material HDPE
Model 9770
Product Type Industrial Safety Helmet
View More Specs

Products 1
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17564106
View Documents
uvex
9770030
White
420.00 €
Pack of 25
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
Description

Description

  • Lightweight helmet in a sporty design
  • Helmet shell fully closed for use by electricians
  • Tested and certified in accordance with EN 397 and EN 50365 (1000 VAC)
  • Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
  • Suspension harness with rotating size adjusters for individual width adjustment
Specifications

Specifications

EN 397, EN 50365
HDPE
Industrial Safety Helmet
52 to 61 cm
Not Ventilated
Hard Hats
9770
52 to 61 cm
Long Brim
Short Brim (Cap Style)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.