missing translation for 'onlineSavingsMsg'
Learn More

uvex Pheos Faceguard Mesh Visor and Earmuffs K1P

Uvex pheos faceguard mesh visor and hearing protection.

Brand:  uvex 9790215

78.25 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 17797709

  • 85.40 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Faceshield multi-system
  • Adjustment to head size via wheel ratchet (52 to 64 cm)
  • Infinitely variable folding mechanism
  • Parking position of visor close to the head
  • System with hearing protection, earmuffs K1P with bayonet closure
  • Stainless steel mesh visor with high light transmittance
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black, Clear
52 to 64 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers