missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Hot Start II PCR Master Mixes

Thermo Scientific Phire Hot Start II PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2.

Brand:  Thermo Scientific™ F125S

185.25 EUR valid until 2025-12-31
Use promo code "24357" to get your promotional price.


 View more versions of this product

Product Code. 15361732

  • 201.00 € / 200 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Phire Hot Start II PCR Master Mix is a convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides, and optimized reaction buffer, including MgCl2. Only template and primers need to be added to the PCR reaction.

Phire Green Hot Start II PCR Master Mix also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Hot Start II DNA Polymerase, included in the master mix, is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain that allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

Features

  • No reactivation step with quick hot start
  • Amplify four times faster than with hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplify significantly longer DNA fragments than with any hot start Taq
  • Direct loading on gel with Green format

Applications

  • Hot Start PCR
  • Routine PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 2X Phire Hot Start II PCR Master Mix, 4 x 1.25 mL
• DMSO, 1 x 0.5 mL
• Nuclease-free water, 4 x 1.25 mL

Store at -20°C.
Phire Hot Start II DNA Polymerase
Hot Start PCR Master Mix
Wet or Dry Ice
2X
Built-In Hot Start
Blunt
DNA
Medium
Fast
200 reactions
Cloning, Hot-start PCR
2X
200 Reactions
SuperMix or Master Mix
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.